Bordet Gengou Agar Base, Dehydrated Culture Media
Supplier: HIMEDIA
M175-500G
HIMEM175-500GEA 127 EUR
HIMEM175-500G
Bordet Gengou Agar Base, Dehydrated Culture Media
Media for Microbiology Dehydrated Media for Microbiology
For the detection and isolation of Bordetella pertussis and Bordetella parapertussis.
  • Microbiological culture media
  • Packaged in 500 g poly bottles