Acetobacter Agar Dehydrated Culture Media
Supplier: HIMEDIA
M238-500G
HIMEM238-500GEA 121 EUR
HIMEM238-500G
Acetobacter Agar Dehydrated Culture Media
Media for Microbiology Dehydrated Media for Microbiology
Dehydrated culture media for enrichment, isolation, cultivation, and maintenance of microorganisms.
  • Microbiological culture media
  • Packaged in 500 g poly bottles
Application: Maintenance of glucose-positive Acetobacter species