Campylo Thioglycollate Medium Base Dehydrated Culture Media
Supplier: HIMEDIA
M908-500G
HIMEM908-500GEA 112 EUR
HIMEM908-500G
Campylo Thioglycollate Medium Base Dehydrated Culture Media
Media for Microbiology Dehydrated Media for Microbiology
For the isolation, maintenance, and transport of cultures of Campylobacter species.
  • Microbiological culture media
  • 500 g poly bottles