Automatic burettes, pellet pattern, lateral stopcock and intermediate valve
Supplier: HIRSCHMANN
352016060 352017060 352017560 352036060 352037060 352037560
HIRS352016060EA 285 EUR
HIRS352016060 HIRS352017060 HIRS352017560 HIRS352036060 HIRS352037060 HIRS352037560
Automatic burettes, pellet pattern, lateral stopcock and intermediate valve
Burettes
Automatic burette, Pellet pattern, class B, brown glass, DURAN® with white graduation.
  • Lateral PTFE stopcock or glass key
  • With intermediate valve
Certifications: Compliance with DIN EN ISO 385.