Embedding system, Technovit® 7100
Supplier: HERAEUS KULZER
66009903 64708992 64708996 64708955 64708991
HKUL66009903EA 57.8 EUR
HKUL66009903 HKUL64708992 HKUL64708996 HKUL64708955 HKUL64708991
Embedding system, Technovit® 7100
Tissue Embedders
Technovit 7100 is a plastic embedding system based on HEMA (2-hydroxyethyl methacrylate). The hydrophillic resin is used in medicine, botany, zoology and in the industry for embedding tissues for light microscope studies. The sections can be used for histological staining and enzyme detection.
  • Reproducibility and reliability of the embedding due to the constant, documented quality controls of the individual components
  • Low polymerisation temperature due to Teflon embedding forms
  • Equal hardening of the block, thus uniform and thinnest possible sections
  • In