Snood caps
Supplier: HUBEI ORIENT INTL TRADING
HOIT110174EA 280 EUR
HOIT110174
Snood caps
Disposable Garments
These disposable caps, with hairnet and peak.
  • Made of polypropylene
Weight: 90 g/m² (peak), 35 g/m² (caps), 14 g/m² (hairnet).
Packaging: 100 pieces/bags; 10 bags/case.