Bottles, wide neck, LDPE, with screw cap
Supplier: hünersdorff GmbH
420600
HUEN420600EA 8.1 EUR
HUEN420600
Bottles, wide neck, LDPE, with screw cap
Bottles Plastic Bottles
LDPE, translucent.
  • Fracture-proof and food-safe
  • Resistant to acids, alkalis and other chemicals
  • Suitable for dishwasher and deep freezer
  • Temperature resistant: –20 to +70 °C
Ideal for keeping samples. Can be easily squashed and retain its shape.