Kanamycin sulphate ∼735 μg/mg (from Streptomyces kanamyceticus), white powder

Supplier: MP Biomedicals
Danger

Synonyms: Kanamycin A, Kanamycin Monosulphate, (2R,3S,4S,5R,6R)-2-(Aminomethyl)-6-(((1R,2R,3S,4R,6S)-4,6-diamino-3-(((2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)tetrahydro-2H-pyran-3,4,5-triol sulphate

0215002901 0215002905 0215002925
ICNA0215002901EA 87.5 EUR
ICNA0215002901 ICNA0215002905 ICNA0215002925
Kanamycin sulphate ∼735 μg/mg (from Streptomyces kanamyceticus), white powder
Kanamycin sulphate

Kanamycin Monosulfate is an aminoglycoside bacteriocidal antibiotic which acts as an inhibitor of protein biosynthesis by producing a misreading of the 70s-ribosome.
Kanamycin Monosulfate used in biotechnology applications to inhibit protein synthesis. It is also used as a selection agent for cells transformed with kanamycin B (neoR, kanR) resistance gene.
Mode of Action: Binds to 70S ribosomal subunit; inhibits translocation; elicits miscoding. Antimicrobial spectrum: Gram-negative and Gram-positive bacteria, and mycoplasma.


  • Presentation: White Powder
  • Soluble in water(10-50 mg/mL).
  • Solutions should be filter sterilized. Store in small aliquots at appropriate temperature until use.
  • Storage temperature: Store at Room Temperature (15-30 °C)

Formula: C₁₈H₃₈N₄O₁₅S
Melting Pt: >250 °C (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00070253
CAS Number: 25389-94-0

Order Now

Specification Test Results

Identity Test Passes
Source Streptomyces kanamycetikus
Potency ~735 μg/mg


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR