Citric acid monohydrate, white crystalline powder

Supplier: MP Biomedicals
Warning

Synonyms: 2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid, 2-Hydroxy-1,2,3-propanetricarboxilic acid

02150699.5 0215069901 0215069905
ICNA02150699.5EA 45.6 EUR
ICNA02150699.5 ICNA0215069901 ICNA0215069905
Citric acid monohydrate, white crystalline powder
Citric acid monohydrate

Citric acid acts as an iron chelator. It can support cholesterol/sterol, ubiquinone and isoprenoid biosynthesis in cell culture applications. In animals, citric acid improves the utilization of nutritional calcium. It is a useful buffer component.


Soluble in water, methanol.

Formula: C₆H₈O₇.H₂O
MDL Number: MFCD00149972
CAS Number: 5949-29-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3160681f96abd248f56672a96b72cc5f6e9d89a2&vid=c4db52c32c630d2c9876e60fc3e9e70a117ea0d8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR