Bromophenol blue sodium salt, dark green powder ACS

Supplier: MP Biomedicals

Synonyms: Bromophenol blue sodium, Sodium α-(3,5-dibromo-4-oxo-2,5-cyclohexadienylidene)-α-(3,5-dibromo-4-hydroxyphenyl)toluenesulphonate, Sodium 2,6-dibromo-4-(3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxido-3H-benzo[c][1,2]oxathiol-3-yl)phenolate, Bromophenol blue water soluble, 4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2,6-dibromophenol] S,S-dioxide sodium salt

0215250605
ICNA0215250605EA 30.4 EUR
ICNA0215250605
Bromophenol blue sodium salt, dark green powder ACS
Bromophenol blue sodium salt

Bromophenol blue is a tracking dye for alkaline and neutral buffer systems. It is used as a tracking dye in DNA, RNA (agarose) and protein (polyacrylamide) gel electrophoresis.


  • Visual transition interval: pH 3,0 (yellow) to pH 4,6 (blue)


Bromophenol blue migrates at approximately the same rate as 300-500bp DNA in agarose gel and at the buffer front in protein polyacrylamide gels.


Soluble in water (30 mg/ml - clear, blueish violet solution) or ethanol (7 mg/ml).


Caution: Research Use Only (RUO).

Formula: C₁₉H₉Br₄NaO₅S
MW: 691.95 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00013793
CAS Number: 34725-61-6

Order Now

Specification Test Results

Clarity of Solution Passes
Visual Transition Interval pH 3.0 (yellow) to pH 4.6 (blue)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR