Tobramycin

Supplier: MP Biomedicals

Synonyms: (2S,3R,4S,5S,6R)-4-Amino-2-(((1S,2S,3R,4S,6R)-4,6-diamino-3-(((2R,3R,5S,6R)-3-amino-6-(aminomethyl)-5-hydroxytetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,5-diol

0215692680 0215692690
ICNA0215692680EA 131 EUR
ICNA0215692680 ICNA0215692690
Tobramycin
Tobramycin

Tobramycin is an aminoglycoside antibiotic derived from Streptomyces tenebrarius and used to treat various types of bacteria infections, particularly Gram-negative infections. It is especially effective against species of Pseudomonas. It works by binding to a site on bacterial 30S and 50S ribosome, preventing formation of the 70S complex. As a result, mRNA cannot be translated into protein and cell death ensues. Tobramycin is preferred over gentamicin for Pseudomonas aeruginosa pneumonia due to better lung penetration.


Tobramycin is used in the treatment of superficial infections of the eye, such as bacterial conjunctivitis. Tobramycin is also indicated for gram-negative infections like meningitis in neonates, brucellosis, pelvic inflammatory disease, Yersinia pestis infection (plague). Its used as an antimicrobial.


Mode of action: Binds to 70S ribosomal subunit, inhibits translocation, and hence elicits miscoding.
Spectrum of activity: Gram negative bacteria.


Soluble in water (50 mg/ml).

Formula: C₁₈H₃₇N₅O₉
MDL Number: MFCD00077885
CAS Number: 32986-56-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR