Tannic acid, brown powder

Supplier: MP Biomedicals

Synonyms: Gallotannin, Tannin

0219027583 0219027590 0219027591
ICNA0219027583EA 58.3 EUR
ICNA0219027583 ICNA0219027590 ICNA0219027591
Tannic acid, brown powder
Tannic acid

Tannic Acid is used for enzyme immobilization and protein adsorption


Tannic acid is used for enzyme immobilisation and protein adsorption.


Soluble in water (2 g/10 ml - clear solution), glycerol, alcohol, acetone. Practically insoluble in benzene, chloroform, ether, petroleum ether, carbondisulphide, carbon tetrachloride. Also soluble in ethanol (100 mg/ml - yellow to brown solution).


Caution: Research Use Only (RUO).

Formula: C₇₆H₅₂O₄₆
Storage Temperature: Ambient
MDL Number: MFCD00066397
CAS Number: 1401-55-4

Order Now

Specification Test Results

Identity Test Passes
Moisture ≤12%
FTIR Conforms to Standard


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR