IPTG (isopropyl-β-D-thiogalactopyranoside) ≥99% for molecular biology

Supplier: MP Biomedicals
Warning

Synonyms: IPTG, (2R,3R,4S,5R,6S)-2-(Hydroxymethyl)-6-(isopropylthio)tetrahydro-2H-pyran-3,4,5-triol, Isopropyl Beta-D-1-thiogalactopyranoside

0219402983 0219402990 0219402980 0219402991 0219402994 0219402995
ICNA0219402983EA 44.9 EUR
ICNA0219402983 ICNA0219402990 ICNA0219402980 ICNA0219402991 ICNA0219402994 ICNA0219402995
IPTG (isopropyl-β-D-thiogalactopyranoside) ≥99% for molecular biology
IPTG (Isopropyl-β-D-thiogalactopyranoside)

Cell culture media component for use in molecular genetics. β-Galactoside inducer.


Storage temperature: -20°C


pH: 5.0 - 7.0 (4% aq soln)

Formula: C₉H₁₈O₅S
MW: 238.31 g/mol
Boiling Pt: 402
Melting Pt: 109.5…110.5 °C
Storage Temperature: Freezer
MDL Number: MFCD00063273
CAS Number: 367-93-1
EINECS: 206-703-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR