Dehydrated culture media, potato dextrose agar, MP Biomedicals
Supplier: MP Biomedicals
091008517
ICNA091008517EA 218 EUR
ICNA091008517
Dehydrated culture media, potato dextrose agar, MP Biomedicals
Media for Microbiology Dehydrated Media for Microbiology
Potato dextrose agar (PDA) is suited for the cultivation of yeasts and moulds from dairy products and other foods. It may also be used to induce sporulation in many fungi.
Certifications: This medium is recommended by the Association of Public Health (APHA).