Dulbecco's Modification of Eagle's Medium, MP Biomedicals
Supplier: MP Biomedicals
091233454
ICNA091233454EA 51.1 EUR
ICNA091233454
Dulbecco's Modification of Eagle's Medium, MP Biomedicals
Cell Culture Media
Dulbecco's Modification of Eagle's Medium with 20 mM HEPES and 4,5 g Dextrose/liter, without L-glutamine or Sodium bicarbonate is ideally suited for supporting and maintaining a vast range of mammalian cell types.
  • Most widely used modification of Eagle's Basal Medium (BME)
  • Contains four times greater concentration of amino acids, vitamins and supplementary components
  • For optimal culturing of other cell types, a modification of 4500 mg/L glucose is available
The original formulation calls for 1000 mg/L of glucose, which was first employed to support the polyoma virus in primary and secondary embryonic mouse cultures.