Rotary evaporators, RV 3 V
Supplier: IKA
10003324 10003328 10003331
IKAA10003324EA 2971 EUR
IKAA10003324 IKAA10003326 IKAA10003328 IKAA10003329 IKAA10003331 IKAA10003332 IKAA10003327 IKAA10003330 IKAA10003333
Rotary evaporators, RV 3 V
Evaporators Rotary Evaporators
The RV 3 rotary evaporator is the ideal entry-level model from the IKA rotary evaporator portfolio. It is suitable for a multitude of uses in the chemical, pharmaceutical, and biotechnology industries, in research and development, in manufacturing or quality assurance, in laboratories, and in plant construction. Thanks to specially designed glass guides, the condenser makes extremely efficient use of the 1500 cm² cooling surface and is space saving.
  • Water heating bath has digital temperature display and carrying handles
  • Mechanical lift end-point safety