Oils for rotational viscometers, ROTAVISC
Supplier: IKA
25000397 25000398 25000399 25000434 25000435 25000436 25000437 25000466 25000467 25000468 25000469
IKAA25000397EA 154 EUR
IKAA25000397 IKAA25000398 IKAA25000399 IKAA25000434 IKAA25000435 IKAA25000436 IKAA25000437 IKAA25000466 IKAA25000467 IKAA25000468 IKAA25000469
Oils for rotational viscometers, ROTAVISC
Lubricants
These certified standard silicone oils can be used to check the measurement accuracy of ROTAVISC.