tempmate.®-GS2 Single Use GSM Data Logger
Supplier: tempmate
G2T-000
IMECG2T-000EA 119 EUR
IMECG2T-000
tempmate.®-GS2 Single Use GSM Data Logger
Data Loggers
Single use GSM data logger for real-time measurement of temperature, humidity and light. GSM tracking and LTE 4G connectivity for smart logistics worldwide.
  • Choose from 3 different devices to find the right model for your application
  • The simple configuration offers the possibility to generate individual reports for your transport routes at up to 3 device starts
  • Live monitoring with free tempmate.®-cloud platform for total transparency in your supply chain
  • Reliable recording of extremely high and low temperatures
  • The integrated USB port allows the
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Interface
Protection class IP 65
Weight 130 g
Data memory - No. of readings Unlimited cloud storage