Lithium dodecyl sulphate 98+% (by acidimetry)

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Dodecyl sulphuric acid lithium salt, Dodecyl sulphate lithium salt, Lithium lauryl sulphate, Lauryl sulphate lithium salt

J32816.22 J32816.14 J32816.A1
J32816.22EA 252 EUR
J32816.22 J32816.14 J32816.A1
Lithium dodecyl sulphate 98+% (by acidimetry)
Lithium dodecyl sulphate
Formula: C₁₂H₂₅LiO₄S
Storage Temperature: Ambient
MDL Number: MFCD00007467
CAS Number: 2044-56-6
EINECS: 218-058-2
UN: 1325
ADR: 4.1,III

Order Now

Specification Test Results

Form White crystalline powder
Assay (unspecified) ≥98% (acidity)
Assay (unspecified) Absorbance (230 nm, 3%, 1 cm) ≤ 0.2
Assay (unspecified) Absorbance (280 nm, 3%, 1 cm) ≤ 0.1
Assay (GC) (C12) ≥98%
Chloride content ≤ 0.01%
Loss on Drying ≤ 1%
Trace Metal: Copper (Cu) ≤ 5 ppm
Trace Metal: Iron (Fe) ≤ 1 ppm
Trace Metal: Lead (Pb) ≤ 5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR