Clofibrate ≥95%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Ethyl 2-(4-chlorophenoxy)-2-methylpropanoate

J60342.03 J60342.14 J60342.06
J60342.03EA 19.3 EUR
J60342.03 J60342.14 J60342.06
Clofibrate ≥95%
Clofibrate

Clofibrate is a ligand agonist for PPARα (peroxisome proliferator-activated receptor α), or an inducer of cytochromes P450’s CYP4A1 and CYP4A3.


  • Applications: Peroxisome proliferator-activated receptor-a agonist. Induces apoptosis in hepatoma cells


Purity: 95+% CAS No.: 637-07-0 Molecular Formula: C12H15ClO3 Formula Weight: 242.70 Storage Temperature: 2°C to 8°C Flash Point: 113°(235°F) Physical Form: Liquid Density: 1.14 Refractive Index: 1.5 Appearance: Clear Boiling Point: 154-156° Beilstein Reference: 1913459 UN No.: UN3082 MDL No.: MFCD00000615


A lipid-lowering agent used for controlling the high cholesterol and triacylglyceride level in the blood. It increases lipoprotein lipase activity to promote the conversion of VLDL to LDL, and hence reduce the level of VLDL. It can increase the level of HDL.

Formula: C₁₂H₁₅ClO₃
Boiling Pt: 154…156 °C
Density: 1.14
Flash Pt: 113 °C (235 °F)
MDL Number: MFCD00000615
CAS Number: 637-07-0
EINECS: 211-277-4
ADR: 9,III
Merck Index: 14,2377

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR