Thymidine-5'-monophosphate disodium salt

Supplier: Thermo Fisher Scientific

Synonyms: 5′-Thymidylic acid disodium salt, TMP disodium salt, Sodium ((2R,3S,5R)-3-hydroxy-5-(5-methyl-2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)tetrahydrofuran-2-yl)methyl phosphate, dTMP disodium salt, Thymidine 5′-monophosphate disodium salt hydrate

J60747.MC J60747.MD J60747.03
J60747.MCEA 46.9 EUR
J60747.MC J60747.MD J60747.03
Thymidine-5'-monophosphate disodium salt
Thymidine-5'-monophosphate disodium salt

CAS No.: 33430-62-5 Molecular Formula: C10H13N2Na2O8P Formula Weight: 366.18 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White Solubility: Soluble in water Beilstein Reference: 4086927 MDL No.: MFCD09039259

Formula: C₁₀H₁₃N₂Na₂O₈P
Storage Temperature: Refrigerator
MDL Number: MFCD09039259
CAS Number: 33430-62-5
EINECS: 251-518-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR