Histamine (free base)

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 2-(1H-Imidazol-4-yl)ethan-1-amine, Histamine

J61727.03
J61727.03EA 94.8 EUR
J61727.03
Histamine (free base)
Histamine (free base)
Formula: C₅H₉N₃
Storage Temperature: Refrigerator
MDL Number: MFCD00005210
CAS Number: 51-45-6
EINECS: 200-100-6
UN: 2811
ADR: 6.1,III
Merck Index: 14,4719

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR