Polymyxin B sulphate, Cell Culture Grade

Supplier: Thermo Fisher Scientific

J61763.03 J61763.06
J61763.03EA 115 EUR
J61763.03 J61763.06
Polymyxin B sulphate, Cell Culture Grade
Polymyxin B sulphate

Antibiotic for Gram-negative bacteria


  • White to pale yellow powder


Soluble in water

Formula: C₅₅H₉₆N₁₆O₁₃·2H₂SO₄
Storage Temperature: Refrigerator
MDL Number: MFCD00131991
CAS Number: 1405-20-5
Merck Index: 14,7573

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR