Magnesium chloride hexahydrate cell culture reagent

Supplier: Thermo Fisher Scientific

Synonyms: Magnesium dichloride

J62575.30 J62575.36 J62575.A1
J62575.30EA 65.6 EUR
J62575.30 J62575.36 J62575.A1
Magnesium chloride hexahydrate cell culture reagent
Magnesium chloride hexahydrate

CAS No.: 7791-18-6 Molecular Formula: MgCl2.6H2O Formula Weight: 203.30 (95.22anhy) Physical Form: Powder Appearance: White Odour: Odourless Merck Reference: 14,5662 MDL No.: MFCD00149781

Formula: MgCl₂·6H₂O
Boiling Pt: 1412 °C (1013 hPa)
Melting Pt: 117 °C
Density: 1.57 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149781
CAS Number: 7791-18-6
EINECS: 232-094-6
Merck Index: 14,5662

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR