Erythrosin B, powder, spirit soluble

Supplier: Thermo Fisher Scientific

Synonyms: Tetraiodofluorescein, Solvent Red 140, C.I. 45430:2

J62619.06 J62619.14 J62619.22
J62619.06EA 34.5 EUR
J62619.06 J62619.14 J62619.22
Erythrosin B, powder, spirit soluble
Erythrosin B
Formula: C₂₀H₈I₄O₅
Storage Temperature: Ambient
MDL Number: MFCD00005044
CAS Number: 15905-32-5
EINECS: 240-046-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR