Orange G for electrophoresis

Supplier: Thermo Fisher Scientific

Synonyms: Disodium 7-hydroxy-8-phenylazonaphthalene-1,3-disulphonate, Wool Orange 2G, Orange G sodium salt, CI 16230, Acid Orange 10

J62743.14 J62743.22 J62743.A1
J62743.14EA 22.1 EUR
J62743.14 J62743.22 J62743.A1
Orange G for electrophoresis
Orange G

CAS No.: 1936-15-8 Molecular Formula: C16H10N2Na2O7S Formula Weight: 452.38 Physical Form: Powder Appearance: Red to orange Solubility: Soluble in water or methyl cellosolve. Slightly soluble in ethanol Beilstein Reference: 4120705 MDL No.: MFCD00012457


Collagen stain for connective tissue

Formula: C₁₆H₁₀N₂Na₂O₇S₂
Melting Pt: 141 °C
Density: 0.45…0.47 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012457
CAS Number: 1936-15-8
EINECS: 217-705-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR