2'-Deoxy-5-methylcytidine 99%

Supplier: Thermo Fisher Scientific

Synonyms: 4-Amino-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-methylpyrimidin-2(1H)-one, 5-mdC, 5-Methyl-2'-deoxycytidine, 1-(2-Deoxy-beta-D-ribofuranosyl)-5-methylcytosine

J65209.03
J65209.03EA 84.4 EUR
J65209.03
2'-Deoxy-5-methylcytidine 99%
2'-Deoxy-5-methylcytidine

Purity: 99% CAS No.: 838-07-3 Molecular Formula: C10H15N3O4 Formula Weight: 241.24 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White to off-white MDL No.: MFCD00006549

Formula: C₁₀H₁₅N₃O₄
MDL Number: MFCD00006549
CAS Number: 838-07-3
EINECS: 212-655-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR