Conductivity cell
Supplier: Cole-Parmer
99959-74
JENW99959-74EA 564.84 EUR
JENW99959-74
Conductivity cell
Electrodes Conductivity Sensors
This probe is manufactured from PVC and stainless steel and is designed for the measurement of purified water.