Safety cans for liquid disposal
Supplier: JUSTRITE MANUFACTURING
12751 12754 14762Z 14765Z
JUSU12751EA 214 EUR
JUSU12751 JUSU12754 119-0280 119-0281
Safety cans for liquid disposal
Safety Cans
HDPE (red or white), with stainless steel hardware.
  • Chemical resistance and leak protection
  • Broad-base design provides stability and resists accidental tip over
  • Fixed position on handle allows lid to stay open for easy pouring
Liquid disposal safety cans are the safe, complaint way to dispose liquid flammable or corrosive. Spring-loaded sealed cap, normally closed, automatically vents between 3 and 5 psig. Safety can is equipped with a chemically resistant stainless steel flame arrester guards against outside ignition. Carbon insert embedded into theMore Product Information