Acetonitrile ≥99.5% for HPLC

Supplier: VWR Chemicals
Danger

Synonyms: Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile

K981-1LEA 415 EUR
K981-1L K981-4L
Acetonitrile ≥99.5% for HPLC
Acetonitrile

  • High purity, functionally tested reagents suitable for HPLC and mass spectrometry.
  • Low UV absorbing and low particulate count to reduce background.

Formula: H₃CCN
Boiling Pt: 81.6 °C (1013 hPa)
Melting Pt: –45.7 °C
Density: 0.782 g/cm³ (20 °C)
Flash Pt: 2 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II
REACH: 01-2119471307-38
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Abs.@220nm <2
Abs.@254nm <0.5
Assay >99.5 %
Liquid Chromatography Suitability Pass
Residue After Evaporation <0.005 %
Titrable Acid <8 meq/g
Titrable Base <0.6 meq/g
Water (KF) <0.3 %

SPECIFICATIONS

Environmentally Preferable Safer Human and Environmental Health
Environmental certification EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR