Beta radiation shields
Supplier: KARTELL
572
KART572EA 547 EUR
KART572
Beta radiation shields
Shields Radiation Shields
Acrylic protection shield, transparent, thickness: 9 mm.
  • Provides protection from radiations while using beta emitting isotopes, including P32
  • Free standing, porable and stable, fitted with non skid feet