Graduated pipettes, Qualicolor, colour coded
Supplier: KAVALIER GLASS
N632434116312 632434116314 632434116516 632434116618 632434116719 N632434116722 632434116723 632434116725 632434119719 632434119723 632434119618 632434119722 632434119312
KAVAN632434116312EA 18.3 EUR
KAVAN632434116312 KAVA632434116314 KAVA632434116516 KAVA632434116618 KAVA632434116719 KAVAN632434116722 KAVA632434116723 KAVA632434116725 KAVA632434119719 KAVA632434119723 KAVA632434119618 KAVA632434119722 KAVA632434119312
Graduated pipettes, Qualicolor, colour coded
Pipettes Graduated Pipettes
Soda-lime glass, class A.
  • Calibrated to deliver (TD)
  • With or without certificate
  • Not autoclavable at 121 °C
Certifications: DIN 12697, ISO 835.