Blood collection tubes with sodium citrate anticoagulant, vacuum, Vacutest®
Supplier: KIMA SAS DI CHIARIN RENZO
14365 14530 140740 143650 14540 140200 14074 14315 14574 14084 140840
720-2748EA 40 EUR
720-2748 720-2772 720-2771 KIMA143650 720-2773 KIMA140200 KIMA14074 KIMA14315 KIMA14574 KIMA14084 KIMA140840
Blood collection tubes with sodium citrate anticoagulant, vacuum, Vacutest®
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
The test tubes are made from Polyethylene terephthalate (PET). Resistant to mechanical stress, centrifugation, breakage during standard procedures of use and in case of accidental falls. PET is a transparent material which enables the visual inspection of the blood and other biological liquids collected. Labels are made from self-adhesive paper for specific medical use, size 40×20 mm, printed in three colours.
  • Available in different sizes
  • Colours of the closure system are compliant with ISO 6710 and EN 14820 standards for the identification of the tubes
  • 18