Gloves, neoprene-nitrile, multi-layered, Kimtech™ Prizm™
Supplier: KIMBERLY CLARK
99221
KIMB99221EA 0 EUR
KIMB99221
Gloves, neoprene-nitrile, multi-layered, Kimtech™ Prizm™
Gloves
Kimtech™ Prizm™ multi-layered neoprene nitrile gloves, designed for scientists by scientists, are an ideal choice for the most demanding scientific applications. One of the thinnest Type A certified protective gloves, providing the perfect balance between precision, protection, and comfort.
  • Protection against a wide range of chemicals
  • Ultra-fingertip grip, even when wet
  • Certified ergonomic comfort
  • Mechanical breach indicator
  • Latex-free, powder-free safety gloves with a nitrile accelerator-free donning layer
Seamlessly protect people and processes whenMore Product Information