Handheld refractometers, HR series
Supplier: A.KRÜSS Optronic
HR181 HRT32 HRN62 HRN82 HR92 HRO32 HRM18 HRMT18 HR27/100 HRS16 HR146 HR25/800 HR900 HR901 HRKFZ1 HROT32 HRKL32 HRB32-T HRB92-T
635-2601EA 0 EUR
635-2601 635-2603 635-2604 635-2606 635-2607 635-2608 635-2609 635-2610 635-2611 635-2612 635-2613 635-2614 635-2615 635-2616 635-2618 KRUEHROT32 KRUEHRKL32 KRUEHRB32-T KRUEHRB92-T
Handheld refractometers, HR series
Refractometers Handheld Refractometers
HR series models possess various scales and additional functions which ensures the appropriate handheld refractometer for many application areas.
  • Reliability when reading, as the measured value does not first have to be converted
  • Selected models have an automatic temperature compensation which increases measurement precision for measurements which are performed at 10 to 40 °C instead of 20 °C
  • For calibration, distilled water is required, or else a small calibration plate is provided
These refractometers are useful for the determination of mixingMore Product Information