Manual polarimeter
Supplier: A.KRÜSS Optronic
P1000-LED
KRUEP1000-LEDEA 1820 EUR
KRUEP1000-LED
Manual polarimeter
Polarimeters
This manual polarimeter features a high-quality metal stand and a sample chamber.
  • Very low power consumption
  • Reading is done via the eyepiece with two verniers
Suitable for all applications in the laboratory on the principle of partial shade.