Cell culture media, Ham's F14 (F-14 Nutrient Medium)
Supplier: BIOWEST
L0138-500
L0138-500EA 0 EUR
L0138-500
Cell culture media, Ham's F14 (F-14 Nutrient Medium)
Cell Culture Media
Ham's F14 medium was developed from Ham's F12 medium, which was originally developed for the serum-free clonal growth of Chinese Hamster Ovary (CHO) cells, lung cells and mouse L-cells. Ham's F14 contains a double concentration of amino acids compared to the Ham's F12 medium. This product is also supplemented with calcium chloride and ascorbic acid.