1,2-Bis(chlorodimethylsilyl)ethane 90%, tech.

Supplier: Thermo Fisher Scientific
Danger

L02570.14 L02570.22
L02570.14EA 52.6 EUR
L02570.14 L02570.22
1,2-Bis(chlorodimethylsilyl)ethane 90%, tech.
1,2-Bis(chlorodimethylsilyl)ethane

Purity: 90% CAS No.: 13528-93-3 Molecular Formula: C6H16Cl2Si2 Formula Weight: 215.27 Flash Point: 65°(149°F) Melting Point: 34-40° Boiling Point: 198-200° Beilstein Reference: 1920141 UN No.: UN3261 MDL No.: MFCD00000505

Formula: C₆H₁₆Cl₂Si₂
Boiling Pt: 198…200 °C
Melting Pt: 34…40 °C
Flash Pt: 65 °C (149 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000505
CAS Number: 13528-93-3
EINECS: 236-871-0
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR