4-(Methylthio)aniline ≥98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 4-(Methylmercapto)aniline

L04950.06 L04950.14
L04950.06EA 63.2 EUR
L04950.06 L04950.14
4-(Methylthio)aniline ≥98%
4-(Methylthio)aniline

Purity: 98% CAS No.: 104-96-1 Molecular Formula: C7H9NS Formula Weight: 139.23 Flash Point: 110°(230°F) Refractive Index: 1.6390 Odour: Unpleasant Boiling Point: 272-273° Beilstein Reference: 774506 UN No.: 2810 MDL No.: MFCD00007889

Formula: C₇H₉NS
Boiling Pt: 272…273 °C
Melting Pt: 156…158 °C
Density: 1.120
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007889
CAS Number: 104-96-1
EINECS: 203-256-3
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR