Methyl-4-(methylsulphonylmethyl)benzoate 98%

Supplier: Thermo Fisher Scientific

L05657.04 L05657.09
L05657.04EA 75.9 EUR
L05657.04 L05657.09
Methyl-4-(methylsulphonylmethyl)benzoate 98%
Methyl-4-(methylsulphonylmethyl)benzoate

Purity: 98% CAS No.: 160446-22-0 Molecular Formula: C10H12O4S Formula Weight: 228.27 Melting Point: 164-166° Beilstein Reference: 7810971 MDL No.: MFCD00051693

Formula: C₁₀H₁₂O₄S
Melting Pt: 164…166 °C
Storage Temperature: Ambient
MDL Number: MFCD00051693
CAS Number: 160446-22-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR