5-Ethyl-2-furaldehyde ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 5-Ethylfurfural

L06659.03 L06659.06
L06659.03EA 51.1 EUR
L06659.03 L06659.06
5-Ethyl-2-furaldehyde ≥98%
5-Ethyl-2-furaldehyde

Purity: 98% CAS No.: 23074-10-4 Molecular Formula: C7H8O2 Formula Weight: 124.14 Flash Point: 65°(149°F) Refractive Index: 1.5220 Boiling Point: 79-81°/12mm Beilstein Reference: 1982 MDL No.: MFCD00030605

Formula: C₇H₈O₂
MW: 124.14 g/mol
Boiling Pt: 79…81 °C (12 mmHg)
Density: 1.055
Flash Pt: 65 °C (149 °F)
Storage Temperature: Ambient
MDL Number: MFCD00030605
CAS Number: 23074-10-4
EINECS: 245-411-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR