2-Methoxy-3-methylpyrazine ≥99%

Supplier: Thermo Fisher Scientific
Danger

L08067.06 L08067.14
L08067.06EA 56.3 EUR
L08067.06 L08067.14
2-Methoxy-3-methylpyrazine ≥99%
2-Methoxy-3-methylpyrazine

Purity: 99% CAS No.: 2847-30-5 Molecular Formula: C6H8N2O Formula Weight: 124.14 Flash Point: 55°(131°F) Refractive Index: 1.5065 Boiling Point: 159° Beilstein Reference: 878809 UN No.: 1993 MDL No.: MFCD00006127

Formula: C₆H₈N₂O
Boiling Pt: 159 °C
Density: 1.070
Flash Pt: 55 °C (131 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006127
CAS Number: 2847-30-5
EINECS: 220-651-6
ADR: 3,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR