2-Chloro-5-(trifluoromethyl)pyridine ≥97%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 2-Chloro-5-trifluoromethylpyridine

L08207.14 L08207.22
L08207.14EA 79 EUR
L08207.14 L08207.22
2-Chloro-5-(trifluoromethyl)pyridine ≥97%
2-Chloro-5-(trifluoromethyl)pyridine

Purity: 97% CAS No.: 52334-81-3 Molecular Formula: C6H3ClF3N Formula Weight: 181.55 Flash Point: 62°(143°F) Melting Point: 29-33° Boiling Point: 152-153° Beilstein Reference: 3649688 UN No.: 2811 MDL No.: MFCD00042225

Formula: C₆H₃ClF₃N
Boiling Pt: 152…153 °C
Melting Pt: 29…33 °C
Density: 1.417
Flash Pt: 62 °C (143 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042225
CAS Number: 52334-81-3
EINECS: 257-856-5
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR