5-Chloro-1-pentanol 95%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 5-Chloropentan-1-ol

L12088.09 L12088.18
L12088.09EA 98.8 EUR
L12088.09 L12088.18
5-Chloro-1-pentanol 95%
5-Chloro-1-pentanol

Purity: 95% CAS No.: 5259-98-3 Molecular Formula: Cl(CH2)5OH Formula Weight: 122.60 Refractive Index: 1.4540 Boiling Point: 102-103°/8mm MDL No.: MFCD00039556

Formula: C₅H₁₁ClO
Boiling Pt: 102…103 °C (8 mmHg)
Density: 1.060
Storage Temperature: Ambient
MDL Number: MFCD00039556
CAS Number: 5259-98-3
EINECS: 226-067-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR