(1S,2S)-1,2-Bis((R)-2,2-dimethyl-1,3-dioxolan-4-yl)ethane-1,2-diol

Supplier: Thermo Fisher Scientific

Synonyms: 1,2:5,6-Di-O-isopropylidene-D-mannitol, (1S,2S)-1,2-bis[(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]ethane-1,2-diol, (1S,2S)-1,2-BIS((R)-2,2-DIMETHYL-1,3-DIOXOLAN-4-YL)ETHANE-1,2-DIOL

L12127.06 L12127.14
L12127.06EA 54 EUR
L12127.06 L12127.14
(1S,2S)-1,2-Bis((R)-2,2-dimethyl-1,3-dioxolan-4-yl)ethane-1,2-diol
(1S,2S)-1,2-Bis((R)-2,2-dimethyl-1,3-dioxolan-4-yl)ethane-1,2-diol

CAS No.: 1707-77-3 Molecular Formula: C12H22O6 Formula Weight: 262.31 Storage Temperature: 2°C to 8°C Melting Point: 120-122° Beilstein Reference: 83765 Optical Rotation: +6° (c=5 in chloroform) MDL No.: MFCD00003211

Formula: C₁₂H₂₂O₆
Melting Pt: 120…122 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003211
CAS Number: 1707-77-3
EINECS: 216-954-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR