Sodium dihydrogen citrate ≥99%

Supplier: Thermo Fisher Scientific

Synonyms: Sodium 3,4-dicarboxy-3-hydroxybutanoate, Sodium citrate monobasic, mono-Sodium citrate, Citric acid monosodium salt, Monosodium Citrate

L12557.36 L12557.22 L12557.0P
L12557.36EA 60.5 EUR
L12557.36 L12557.22 L12557.0P
Sodium dihydrogen citrate ≥99%
Sodium dihydrogen citrate

Purity: 99% CAS No.: 18996-35-5 Molecular Formula: C6H7NaO7 Formula Weight: 214.11 Melting Point: 211-212° Beilstein Reference: 3920246

Formula: C₆H₇NaO₇
Melting Pt: 211…212 °C
Storage Temperature: Ambient
MDL Number: MFCD00013067
CAS Number: 18996-35-5
EINECS: 242-734-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR