Isopentyl nitrite (mixed isomers ) 97% stabilised

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Isoamyl nitrite

Stabilization: Stabilised with sodium carbonate (Na₂CO₃) 0.2% (anhy)

L13909.AC L13909.AE L13909.AP
L13909.ACEA 28.6 EUR
L13909.AC L13909.AE L13909.AP
Isopentyl nitrite (mixed isomers ) 97% stabilised
Isopentyl nitrite (mixed isomers )

Stabilized with 0.2% anhyd. sodium carbonate


Purity: 97% CAS No.: 110-46-3 Molecular Formula: (CH3)2CHCH2CH2ONO Formula Weight: 117.15 Storage Temperature: 2°C to 8°C Flash Point: -20°(-4°F) Refractive Index: 1.3870 Odour: Sweet Boiling Point: 95-100° Beilstein Reference: 969510 Merck Reference: 14,5123 UN No.: 1113 MDL No.: MFCD00002057

Formula: C₅H₁₁NO₂
Boiling Pt: 95…100 °C
Density: 0.875
Flash Pt: –20 °C (–4 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00002057
CAS Number: 110-46-3
EINECS: 203-770-8
UN: 1113
ADR: 3,II
Merck Index: 14,5123

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR