1-Aminopiperidine 97%

Supplier: Thermo Fisher Scientific
Danger

L14078.06 L14078.14 L14078.22
L14078.06EA 59.1 EUR
L14078.06 L14078.14 L14078.22
1-Aminopiperidine 97%
1-Aminopiperidine

Purity: 97% CAS No.: 2213-43-6 Molecular Formula: C5H12N2 Formula Weight: 100.16 Flash Point: 36°(96°F) Refractive Index: 1.4750 Boiling Point: 145-147° Beilstein Reference: 383565 UN No.: 2733 MDL No.: MFCD00006489

Formula: C₅H₁₂N₂
Boiling Pt: 145…147 °C
Density: 0.928
Flash Pt: 36 °C (96 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006489
CAS Number: 2213-43-6
EINECS: 218-666-8
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR