5-Methylthiazole ≥97%

Supplier: Thermo Fisher Scientific
Warning

L15879.06 L15879.14
L15879.06EA 205 EUR
L15879.06 L15879.14
5-Methylthiazole ≥97%
5-Methylthiazole

Purity: 97% CAS No.: 3581-89-3 Molecular Formula: C4H5NS Formula Weight: 99.16 Flash Point: 42°(107°F) Refractive Index: 1.5280 Odour: Stench Boiling Point: 141-142° Beilstein Reference: 104674 UN No.: 1993 MDL No.: MFCD00005335

Formula: C₄H₅NS
Boiling Pt: 141…142 °C
Melting Pt: –40.4 °C
Density: 1.120
Flash Pt: 42 °C (107 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005335
CAS Number: 3581-89-3
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR