(3-Acryloxypropyl)trimethoxysilane ≥94% stabilised

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 3-(Trimethoxysilyl)propyl acrylate

Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

L16400.06 L16400.14
L16400.06EA 109 EUR
L16400.06 L16400.14
(3-Acryloxypropyl)trimethoxysilane ≥94% stabilised
(3-Acryloxypropyl)trimethoxysilane

Stabilized with 100 ppm BHT


Purity: 94% CAS No.: 4369-14-6 Molecular Formula: C9H18O5Si Formula Weight: 234.33 Flash Point: 123°(253°F) Refractive Index: 1.4290 Boiling Point: 95-100°/3mm Beilstein Reference: 2046320 UN No.: 1760 MDL No.: MFCD00054803

Formula: C₉H₁₈O₅Si
Boiling Pt: 95…100 °C (3 mmHg)
Density: 1.059
Flash Pt: 123 °C (253 °F)
Storage Temperature: Ambient
MDL Number: MFCD00054803
CAS Number: 4369-14-6
EINECS: 419-560-6
UN: 1760
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR